Learn More
Cell Biologics HUMAN OVARIAN TUMOR-ASSOCIATED ENDOTHELIAL CELLS
SDP
Catalog No. NC2738940
Encompass_Preferred
Human Primary Ovarian Microvascular Tumor-Associated Endothelial Cells from Cell Biologics are isolated from human ovarian tumor tissue and grown in gelatin pre-coated tissue culture flask with Cell Biologics’ Complete Growth Medium. Cells at passage 3 are harvested from flasks and cryo-preserved in vials. Each vial contains 0.5x106 cells per ml and is delivered frozen. Human Primary Ovarian Microvascular Tumor-Associated Endothelial Cells are characterized by immunofluorescence staining with antibodies of CD31 (Catalog No. 550389, BD), CD31/PECAM-1 (Catalog No. FAB3567P, R&D), VE-Cadherin (FITC-VE-cadherin, Catalog No. 560411, BD), or use of fluorescence-labeled acetylated low-density lipoprotein (DiI-Ac-LDL) uptake (Catalog No. L-35353, Invitrogen), a functional marker for Tumor-Associated Endothelial Cells. These cells are negative for bacteria, yeast, fungi, and mycoplasma and can be expanded for 3-5 passages at a split ratio of 1:2
Catalog No. NC2738940 Supplier Cell Biologics Supplier No. HC6090FROZEN VIAL
Add to Cart
Add to Cart
Human Primary Ovarian Microvascular Tumor-Associated Endothelial Cells from Cell Biologics are isolated from human ovarian tumor tissue and grown in gelatin pre-coated tissue culture flask with Cell Biologics’ Complete Growth Medium. Cells at passage 3 are harvested from flasks and cryo-preserved in vials. Each vial contains 0.5x106 cells per ml and is delivered frozen. Human Primary Ovarian Microvascular Tumor-Associated Endothelial Cells are characterized by immunofluorescence staining with antibodies of CD31 (Catalog No. 550389, BD), CD31/PECAM-1 (Catalog No. FAB3567P, R&D), VE-Cadherin (FITC-VE-cadherin, Catalog No. 560411, BD), or use of fluorescence-labeled acetylated low-density lipoprotein (DiI-Ac-LDL) uptake (Catalog No. L-35353, Invitrogen), a functional marker for Tumor-Associated Endothelial Cells. These cells are negative for bacteria, yeast, fungi, and mycoplasma and can be expanded for 3-5 passages at a split ratio of 1:2