Learn More
Creative Biolabs Human OVCAR-8 Tumor Cell Line, Tumor Cell Line, OVCAR-8, Ovarian Adenocarcinoma, Epithelial, Adherent morphology, Storage: Liquid nitrogen (-150°C to -190°C), Applications: Cancer research, Drug screening
SDP
Catalog No. NC3706344
Encompass
Human OVCAR-8 Tumor Cell Line, Tumor Cell Line, OVCAR-8, Ovarian Adenocarcinoma, Epithelial, Adherent morphology, Storage: Liquid nitrogen (-150°C to -190°C), Applications: Cancer research, Drug screening
Catalog No. NC3706344 Supplier Creative Biolabs Supplier No. IOCZP305CBLC041425JF5A
Human OVCAR-8 Tumor Cell Line, Tumor Cell Line, OVCAR-8, Ovarian Adenocarcinoma, Epithelial, Adherent morphology, Storage: Liquid nitrogen (-150°C to -190°C), Applications: Cancer research, Drug screening