Learn More
ACROBiosystems Recombinant Protein;Human PD-L2 / B7-DC Protein;HEK293;100UG;PD2,PDL2,PD-L2,Butyrophilin B7-DC,CD273,PDCD1 ligand 2,PDCD1L2,PDCD1LG2,PDL-2,B7DC
Supplier:  ACROBiosystems PD2H5220100UG
Encompass_Preferred
Recombinant Protein;Human PD-L2 / B7-DC Protein;100UG;Human PD-L2, His Tag (PD2-H5220) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession # AAI13679).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.4 kDa. The protein migrates as 34-46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-L2, His Tag (Cat. No. PD2-H5220) at 5 μg/mL ( 100 μl/well ) can bind Human PD-1, Fc Tag (Cat. No. PD1-H5257) with a linear range of 0.02-0.5 μg/mL (QC tested).
Catalog No. 50-101-8432