Learn More
STEMCELL Technologies Human Peripheral Blood Gamma Delta T Cells, Frozen
SDP
Catalog No. NC2832260
Encompass
Gamma delta T cells (TCRγδ+CD3+) are isolated from peripheral blood using negative immunomagnetic separation and cryopreserved in animal component-free CryoStor®CS10 medium (Catalog #07930). Cells are obtained using Institutional Review Board (IRB)-approved consent forms and protocols. Additional documentation, high-resolution HLA typing (Class I and Class II alleles), and viral screening status are available upon request. Acid-citrate-dextrose solution A (ACDA) is added during collection as an anticoagulant.
Catalog No. NC2832260 Supplier STEMCELL Technologies Supplier No. 2000730
Gamma delta T cells (TCRγδ+CD3+) are isolated from peripheral blood using negative immunomagnetic separation and cryopreserved in animal component-free CryoStor®CS10 medium (Catalog #07930). Cells are obtained using Institutional Review Board (IRB)-approved consent forms and protocols. Additional documentation, high-resolution HLA typing (Class I and Class II alleles), and viral screening status are available upon request. Acid-citrate-dextrose solution A (ACDA) is added during collection as an anticoagulant.