Learn More
Abcam Human PreAlbumin ELISA Kit (Transthyretin), 1 x 96 Tests
Supplier:  Abcam AB1088951X96TESTS
Encompass_Preferred
Human PreAlbumin ELISA Kit (Transthyretin) ab108895 is used to quantify Human PreAlbumin with a sensitivity of 7.8 pg/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1737866