Learn More
Angio Proteomie Human Retinal Astrocytes, 1.0 Frozen Vial
SDP
Supplier:  Angio Proteomie CAP0115
Encompass
HRAs (cAP-0115) are isolated from normal human retinal tissue. The cells are shipped in frozen vials (the cells are provided @ passage 2). Astrocyte Growth Medium (AsGM, contains FBS and Growth factor supplements, cAP-29) is recommended for cell culture and these cells have a minimum average population doubling levels > 10 when cultured following the detailed protocol described below).
Catalog No. NC3525628