Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED ON-TARGETplus Human RRAGC (64121) siRNA - Set of 4, Unit Size: 5 nmol
Catalog No. NC3782720
Encompass
This gene encodes a member of the GTR/RAG GTP-binding protein family. The encoded protein is a monomeric guanine nucleotide-binding protein which forms a heterodimer with RRAGA and RRAGB and is primarily localized to the cytoplasm. The encoded protein promotes intracellular localization of the mTOR complex. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Dec 2012].
Catalog No. NC3782720 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. LQ017822010005
This gene encodes a member of the GTR/RAG GTP-binding protein family. The encoded protein is a monomeric guanine nucleotide-binding protein which forms a heterodimer with RRAGA and RRAGB and is primarily localized to the cytoplasm. The encoded protein promotes intracellular localization of the mTOR complex. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Dec 2012].