Learn More
ACROBiosystems Human TL1A-DR3 inhibition Kit (TR-FRET) 500 Tests
Catalog No. NC3740815
Encompass_Preferred
Cost effective - Sufficient quantity at a lower price, accounting for dilution and pipetting losses.Comprehensive validation - Validated with various antibody subtypes and antibody drugs.Simple and fast operation - No complicated washing steps, significantly reducing time.High batch consistency - Strict control over raw materials and finished product quality, ensuring a stable supply.Accurate and reliable results - High sensitivity with minimal matrix effects.High throughput capability - Supports 500 tests, ideal for high-throughput screening.Fast completion - Results in just 1 hour.
Catalog No. NC3740815 Supplier ACROBiosystems Supplier No. FRT02500TESTS
Cost effective - Sufficient quantity at a lower price, accounting for dilution and pipetting losses.Comprehensive validation - Validated with various antibody subtypes and antibody drugs.Simple and fast operation - No complicated washing steps, significantly reducing time.High batch consistency - Strict control over raw materials and finished product quality, ensuring a stable supply.Accurate and reliable results - High sensitivity with minimal matrix effects.High throughput capability - Supports 500 tests, ideal for high-throughput screening.Fast completion - Results in just 1 hour.