Learn More
ACROBiosystems Recombinant Protein;ActiveMax Human VEGF121 Protein, Tag Free;HEK293;50UG;VE1,RP1-261G23.1,MGC70609,MVCD1,MVCD-1,VEGF 165,VEGF-A,VEGFA,VPF,VEGF165,VEGF-165,VEGF121,VEGF120,VEGF186,VEGF164,VEGF110
Catalog No. 501018737
Encompass_Preferred
Recombinant Protein;ActiveMax Human VEGF121 Protein, Tag Free;50UG;ActiveMax Human VEGF121, Tag Free (VE1-H4213) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 147 (Accession # NP_001020541).;This protein carries no "tag". The protein has a calculated MW of 14 kDa (monomer). As a result of glycosylation, the protein migrates as 19 kDa and 20kDa under reducing (R) condition, and 40-45 kDa (homodimer) under non-reducing (NR) condition (SDS-PAGE).;Immobilized ActiveMax Human VEGF121, Tag Free (Cat. No. VE1-H4213) at 2 g/mL can bind Human VEGF R2, Strep Tag (Cat. No. KDR-H5280) with a linear range of 0.15-2.5 g/mL (QC tested).
Catalog No. 50-101-8737 Supplier ACROBiosystems Supplier No. VE1H421350UG
Recombinant Protein;ActiveMax Human VEGF121 Protein, Tag Free;50UG;ActiveMax Human VEGF121, Tag Free (VE1-H4213) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 147 (Accession # NP_001020541).;This protein carries no "tag". The protein has a calculated MW of 14 kDa (monomer). As a result of glycosylation, the protein migrates as 19 kDa and 20kDa under reducing (R) condition, and 40-45 kDa (homodimer) under non-reducing (NR) condition (SDS-PAGE).;Immobilized ActiveMax Human VEGF121, Tag Free (Cat. No. VE1-H4213) at 2 g/mL can bind Human VEGF R2, Strep Tag (Cat. No. KDR-H5280) with a linear range of 0.15-2.5 g/mL (QC tested).