Learn More
ACROBiosystems Biotinylated Human CD3 epsilon&CD3 gamma Heterodimer Protein, His,Avitag™&Tag Free (MALS verified)
Catalog No. 502329179
Encompass_Preferred
Biotinylated Human CD3E&CD3G Heterodimer Protein, His,Avitag&Tag Free (CDG-H82W3) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # NP_000724.1 (CD3E) & AAI13831 (CD3G)).
Catalog No. 50-232-9179 Supplier ACROBiosystems Supplier No. CDGH82W3200UG
Biotinylated Human CD3E&CD3G Heterodimer Protein, His,Avitag&Tag Free (CDG-H82W3) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # NP_000724.1 (CD3E) & AAI13831 (CD3G)).