Learn More
ACROBiosystems Unconjugated Human HLA-A*11:01&B2M Complex Protein (Peptide free, MALS verified)
Catalog No. 502331174
Encompass_Preferred
Unconjugated Human HLA-A*11:01&B2M Complex Protein (HLM-H52H7) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*11:01) & Ile 21 - Met 119 (B2M) (Accession # AAV53343.1 (HLA-A*11:01) & P61769 (B2M)).
Catalog No. 50-233-1174 Supplier ACROBiosystems Supplier No. HLMH52H725UG
Unconjugated Human HLA-A*11:01&B2M Complex Protein (HLM-H52H7) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*11:01) & Ile 21 - Met 119 (B2M) (Accession # AAV53343.1 (HLA-A*11:01) & P61769 (B2M)).