Learn More
ACROBiosystems Biotinylated Human HLA-A*02:01&B2M Complex Protein (Peptide free, MALS verified)
Catalog No. 502332064
Encompass_Preferred
Biotinylated Human HLA-A*02:01&B2M Complex Protein (HLM-H82W3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*02:01) & Ile 21 - Met 119 (B2M) (Accession # AAA59606.1 (HLA-A*02:01) & P61769 (B2M)).
Catalog No. 50-233-2064 Supplier ACROBiosystems Supplier No. HLMH82W325UG
Biotinylated Human HLA-A*02:01&B2M Complex Protein (HLM-H82W3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*02:01) & Ile 21 - Met 119 (B2M) (Accession # AAA59606.1 (HLA-A*02:01) & P61769 (B2M)).