Learn More
ACROBiosystems Human FSH alpha & beta Heterodimer Protein, His Tag
Supplier: ACROBiosystems FSAH52H350UG
Human FSH alpha & beta Heterodimer Protein, His Tag (FSA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Ser 116 (FSH alpha) & Asn 19 - Glu 129 (FSH beta) (Accession # P01215 (FSH alpha) & P01225 (FSH beta)).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.