Learn More
Abcam Human 14-3-3 tau ELISA Kit, 1 x 96 Tests
Catalog No. 502672437
Encompass_Preferred
Human 14-3-3 tau ELISA Kit is a single-wash 90-min Simplestep used to quantify Human 14-3-3 tau with a sensitivity of 6.1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2437 Supplier Abcam Supplier No. AB2465431X96TESTS
Human 14-3-3 tau ELISA Kit is a single-wash 90-min Simplestep used to quantify Human 14-3-3 tau with a sensitivity of 6.1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement