Learn More
Raybiotech Inc Human 15-PGDH/HPGD ELISA Kit
Supplier:  Raybiotech Inc ELH15PGDH1
Encompass_Preferred
For Cell Culture Supernatants. Sensitivity: 0.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human 15-PGDH.
Catalog No. 50-194-8933