Learn More
Sino Biological Human 15-PGDH Matched ELISA Antibody Pair Set 5 Plates
Supplier: Sino Biological SEK112055
The Human HPGD/15-PGDH ELISA Pair Set is for the quantitative determination of Human HPGD/15-PGDH.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human HPGD/15-PGDH coated on a 96-well plate. Standards and samples are added to the wells, and any Human HPGD/15-PGDH present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human HPGD/15-PGDH monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human HPGD/15-PGDH present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.