Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human 17-beta-HSD1/HSD17B1 ELISA Kit

Catalog No. EH2RBX10
Encompass_Preferred
Change view
Click to view available options
10 x 96 Tests; HRP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EH2RBX10 10 x 96 Tests; HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH2RBX10 Supplier Invitrogen™ Supplier No. EH2RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human 17-beta-HSD1/HSD17B1 quantitates human 17-beta-HSD1/HSD17B1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human 17-beta-HSD1/HSD17B1.

HSD17B1 is favors the reduction of estrogens and androgens. It also has 20-alpha-HSD activity. It uses preferentially NADH.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14061
Assay Range 0.9-200 ng/mL
Assay Sensitivity 0.9 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 17-beta-dehydrogenase 1; 17beta-HSD; 17-beta-HSD 1; 17-beta-hydroxysteroid dehydrogenase type 1; 17beta-hydroxysteroid dehydrogenase type 1; 17BHD1; 17HSDB1; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; E17KSR; E2DH; Edh17b1; EDH17B2; EDHB17; Estradiol 17-beta-dehydrogenase 1; estradiol 17-beta-dehydrogenase-1; HSD17; HSD17B1; Hsd17ba; hydroxysteroid (17-beta) dehydrogenase 1; hydroxysteroid (17-beta) dehydrogenase 1 isoform; hydroxysteroid 17-beta dehydrogenase 1; Hydroxysteroid dehydrogenase 17 beta, type 1; LOW QUALITY PROTEIN: estradiol 17-beta-dehydrogenase 1; placental 17-beta-hydroxysteroid dehydrogenase; SDR28C1; Short chain dehydrogenase/reductase family 28C member 1; short chain dehydrogenase/reductase family 28CE, member 117-beta-dehydrogenase 1; 17beta-HSD; 17-beta-HSD 1; 17-beta-hydroxysteroid dehydrogenase type 1; 17beta-hydroxysteroid dehydrogenase type 1; 17BHD1; 17HSDB1; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; E17KSR; E2DH; Edh17b1; EDH17B2; EDHB17; Estradiol 17-beta-dehydrogenase 1; estradiol 17-beta-dehydrogenase-1; HSD17; HSD17B1; Hsd17ba; hydroxysteroid (17-beta) dehydrogenase 1; hydroxysteroid (17-beta) dehydrogenase 1 isoform; hydroxysteroid 17-beta dehydrogenase 1; Hydroxysteroid dehydrogenase 17 beta, type 1; LOW QUALITY PROTEIN: estradiol 17-beta-dehydrogenase 1; placental 17-beta-hydroxysteroid dehydrogenase; SDR28C1; Short chain dehydrogenase/reductase family 28C member 1; short chain dehydrogenase/reductase family 28CE, member 1;
Gene ID (Entrez) 3292
Gene Symbol HSD17B1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target HSD17B1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.