Learn More
Biomatik Corporation Human Α2 Macroglobulin, Α2 Mg Elisa Kit
Supplier:  Biomatik Corporation EKC31359
Encompass_Preferred
Human Α2 Macroglobulin, Α2 Mg Elisa Kit for detecting Alpha-2-Macroglobulin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-20ug/ml. Sensitivity: 1.25ug/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P01023; A2M. Target Synonyms: A2m; A2MG_HUMAN; Alpha 2 M; Alpha 2M; Alpha-2-M; Alpha-2-macroglobulin; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5; CPAMD5; DKFZp779B086; FWP007; S863 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9190