Learn More
Raybiotech Inc Human 2B4 (CD244) ELISA Kit
Supplier:  Raybiotech Inc ELH2B41
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.05 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human 2B4/CD244/SLAMF4.
Catalog No. 50-194-8935