Learn More
Santa Cruz Biotechnology HUMAN 3T3 CELL LYSATE
Catalog No. NC9649113
Encompass
NIH/3T3 normal embryo fibroblast Whole Cell Lysates for Western Blotting Applications 500microgram protein in 200microliter SDS-PAGE Western blotting buffer should be stored at-20degreeC
Catalog No. NC9649113 Supplier Santa Cruz Biotechnology Supplier No. SC2210
NIH/3T3 normal embryo fibroblast Whole Cell Lysates for Western Blotting Applications 500microgram protein in 200microliter SDS-PAGE Western blotting buffer should be stored at-20degreeC