Learn More
ACROBiosystems Human 4-1BB Ligand 2.5mg
Catalog No. 502534459
Encompass_Preferred
ActiveMax Human 4-1BB Ligand / TNFSF9 (71-254) uBeads premium grade (for cells) are uniform superparamagnetic beads of 5.5 µm with streptavidin on its surface and coupled with biotinylated Human 4-1BB Ligand / TNFSF9 (71-254) Protein expressed from human 293 cells (HEK293) and contains AA Arg 71 - Glu 254 (Accession # P41273-1 ).
Catalog No. 50-253-4459 Supplier ACROBiosystems Supplier No. MBSC0122.5MG
ActiveMax Human 4-1BB Ligand / TNFSF9 (71-254) uBeads premium grade (for cells) are uniform superparamagnetic beads of 5.5 µm with streptavidin on its surface and coupled with biotinylated Human 4-1BB Ligand / TNFSF9 (71-254) Protein expressed from human 293 cells (HEK293) and contains AA Arg 71 - Glu 254 (Accession # P41273-1 ).