Learn More
ACROBiosystems Human 4-1BB Ligand 25ug
Catalog No. 502535538
Encompass_Preferred
Biotinylated Human 4-1BB (71-254) Ligand FcAvitag premium grade (41L-H82Fa) is expressed from human 293 cells (HEK293). It contains AA Arg 71 - Glu 254 (Accession # P41273-1).
Catalog No. 50-253-5538 Supplier ACROBiosystems Supplier No. 41LH82FA25UG
Biotinylated Human 4-1BB (71-254) Ligand FcAvitag premium grade (41L-H82Fa) is expressed from human 293 cells (HEK293). It contains AA Arg 71 - Glu 254 (Accession # P41273-1).