Learn More
ACROBiosystems ACROBiosystems Human 4-1BB / TNFRSF9 (87-186) Protein, Fc Tag (MALS verified)
Catalog No. 502106400
Encompass_Preferred
Recombinant Protein;100UG;Human 4-1BB (87-186), Fc Tag (41B-H5253) is expressed from human 293 cells (HEK293). It contains AA Asp 87 - Gln 186 (Accession # Q07011-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 37.1 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6400 Supplier ACROBiosystems Supplier No. 41BH5253100UG
Recombinant Protein;100UG;Human 4-1BB (87-186), Fc Tag (41B-H5253) is expressed from human 293 cells (HEK293). It contains AA Asp 87 - Gln 186 (Accession # Q07011-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 37.1 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.