Learn More
ACROBiosystems Human 4-1BB / TNFRSF9 Protein, His Tag (MALS verified)
Catalog No. 502054954
Encompass_Preferred
Recombinant Protein;100UG;Human 4-1BB, His Tag (41B-H52Hc) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Gln 186 (Accession # Q07011-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.2 kDa. The protein migrates as 27-35 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Measured by its binding ability in a functional ELISA. Immobilized Human 4-1BB Ligand (71-254), Fc Tag, active trimer (Cat. No. 41L-H5265) at 1 μg/mL (100 μL/well) can bind Human 4-1BB, His Tag (Cat. No. 41B-H52Hc) with a linear range of 1-20 ng/mL (QC tested).;FACS analysis shows that Human 4‑1BB, His Tag (Cat.No. 41B-H52Hc) can bind to 4-1BBL cells surface 4-1BB Ligand. The concentration of Human 4-1BB is 1 μg/mL.
Catalog No. 50-205-4954 Supplier ACROBiosystems Supplier No. 41BH52HC100UG
Recombinant Protein;100UG;Human 4-1BB, His Tag (41B-H52Hc) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Gln 186 (Accession # Q07011-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.2 kDa. The protein migrates as 27-35 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Measured by its binding ability in a functional ELISA. Immobilized Human 4-1BB Ligand (71-254), Fc Tag, active trimer (Cat. No. 41L-H5265) at 1 μg/mL (100 μL/well) can bind Human 4-1BB, His Tag (Cat. No. 41B-H52Hc) with a linear range of 1-20 ng/mL (QC tested).;FACS analysis shows that Human 4‑1BB, His Tag (Cat.No. 41B-H52Hc) can bind to 4-1BBL cells surface 4-1BB Ligand. The concentration of Human 4-1BB is 1 μg/mL.