Learn More
Biomatik Corporation Human 4-Hydroxyphenylpyruvate Dioxygenase (HPD) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10512
Encompass_Preferred
Human 4-Hydroxyphenylpyruvate Dioxygenase (HPD) Wide-range ELISA Kit for detecting HPD in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.27ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32754. Gene ID: 3242. Target Synonyms: PPD; 4-HPPD; 4HPPD; GLOD3; Glyoxalase Domain Containing 3; 4-hydroxyphenylpyruvic acid oxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7433