Learn More
Biomatik Corporation Human 5',3'-Nucleotidase, Cytosolic (NT5C) ELISA Kit
Supplier:  Biomatik Corporation EKU10600
Encompass_Preferred
Human 5',3'-Nucleotidase, Cytosolic (NT5C) ELISA Kit for detecting NT5C in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8TCD5. Gene ID: 30833. Target Synonyms: DNT; DNT1; P5N2; PN-I; PN-II; UMPH2; cdN; 5'Nucleotidase,Deoxy (Pyrimidine) Cytosolic Type C; Uridine 5-Prime Monophosphate Hydrolase 2; 5' (3')-deoxyribonucleotidase, cytosolic. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7051