Learn More
BIOHIPPO Human 5HTR5A Microsample
SDP
Catalog No. 5000314830
Encompass
Human 5HTR5A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of 5HTR5A in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 064 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human 5HTR5A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human 5HTR5A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human 5HTR5A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14830 Supplier BIOHIPPO Supplier No. BHE15210633
Add to Cart
Add to Cart
Human 5HTR5A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of 5HTR5A in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 064 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human 5HTR5A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human 5HTR5A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human 5HTR5A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color