Learn More
Biomatik Corporation Human 5-Hydroxytryptamine Receptor 1A (HTR1A) ELISA Kit
Supplier:  Biomatik Corporation EKU02028
Encompass_Preferred
Human 5-Hydroxytryptamine Receptor 1A (HTR1A) ELISA Kit for detecting HTR1A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08908. Gene ID: 3350. Target Synonyms: 5HTR1A; 5HTR1-A; 5-HTR1-A; ADRB2RL1; ADRBRL1; Serotonin Receptor 1A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7699