Learn More
Biomatik Corporation Human 5-Methyltetrahydrofolate Homocysteine Methyltransferase (MTR) ELISA Kit
Supplier:  Biomatik Corporation EKU02033
Encompass_Preferred
Human 5-Methyltetrahydrofolate Homocysteine Methyltransferase (MTR) ELISA Kit for detecting MTR in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.278ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99707. Gene ID: 4548. Target Synonyms: CblG; MS; MeSe; MetH Methionine Synthase; Vitamin-B12 dependent methionine synthase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6718