Learn More
Sino Biological Human 5T4 / TPBG Protein (ECD, His Tag) 100ug
Supplier:  Sino Biological 19845H08H1100
Encompass_Preferred
A DNA sequence encoding the Human TPBG (NP_006661.1) (Met1-Ser355) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6405