Learn More
American Research Products Inc Human 6 hydroXymelatonin sulfa
Catalog No. 5000327974
Encompass
Human 6-hydroXymelatonin sulfate (6HMS) ELISA kit from CUSABIO Detection Range 10 pg/mL- 400 pg/mL Sensitivity 10 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27974 Supplier American Research Products Inc Supplier No. CSBEQ027870HU
Human 6-hydroXymelatonin sulfate (6HMS) ELISA kit from CUSABIO Detection Range 10 pg/mL- 400 pg/mL Sensitivity 10 pg/mL Sample serum plasma tissue homogenates Method Competitive