Learn More
Biomatik Corporation Human A Disintegrin And Metalloprotease 22 (ADAM22) ELISA Kit
Supplier:  Biomatik Corporation EKU02048
Encompass_Preferred
Human A Disintegrin And Metalloprotease 22 (ADAM22) ELISA Kit for detecting ADAM22 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9P0K1. Gene ID: 53616. Target Synonyms: MDC2; Disintegrin And Metalloproteinase Domain-Containing Protein 22; Metalloproteinase-Like,Disintegrin-Like,And Cysteine-Rich Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9157