Learn More
American Research Products Inc Human A Disintegrin And Metall
Catalog No. 5000329524
Encompass
Human A Disintegrin And Metalloprotease 12 ADAM12 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma urine cell culture supernates tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29524 Supplier American Research Products Inc Supplier No. CSBE13163H
Human A Disintegrin And Metalloprotease 12 ADAM12 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma urine cell culture supernates tissue homogenates cell lysates Method Sandwich