Learn More
Biomatik Corporation Human A Disintegrin And Metalloprotease 19 (ADAM19) ELISA Kit
Supplier:  Biomatik Corporation EKU02047
Encompass_Preferred
Human A Disintegrin And Metalloprotease 19 (ADAM19) ELISA Kit for detecting ADAM19 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.108ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H013. Gene ID: 8728. Target Synonyms: FKSG34; MADDAM; MLTNB; meltrin beta; Metalloprotease and disintegrin dendritic antigen marker; Disintegrin and metalloproteinase domain-containing protein 19. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7639