Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human A-GHRL ELISA Kit

Catalog No. EEL181
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL181 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL181 Supplier Invitrogen™ Supplier No. EEL181
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human Acylated Ghrelin (A GHRL) ELISA quantitates A GHRL in serum, plasma and other biological fluids. Principle of the method The Human Acylated Ghrelin (A GHRL) solid phase sandwich ELISA (enzyme linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target specific antibody has been pre coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Acylated ghrelin is a hormone primarily produced by the stomach and released into the bloodstream. It plays a significant role in regulating appetite and energy balance. Functionally, acylated ghrelin acts as an orexigenic hormone, meaning it stimulates appetite and promotes food intake. It interacts with specific receptors in the brain, particularly the growth hormone secretagogue receptor 1a (GHSR1a), to increase hunger signals and enhance food-seeking behaviors. Acylated ghrelin also influences energy homeostasis by increasing fat storage and reducing energy expenditure. Structurally, acylated ghrelin is a peptide hormone consisting of 28 amino acids. It undergoes a post-translational modification process called acylation, where a fatty acid (usually octanoic acid) is attached to the third amino acid residue (serine) in the peptide chain. This acylation is crucial for the hormone's biological activity and stability. Localization-wise, acylated ghrelin is primarily produced by the X/A-like cells in the gastric mucosa. After secretion into the bloodstream, it acts on various target tissues, including the hypothalamus, pituitary gland, and adipose tissue. The hypothalamus is particularly important for regulating appetite and energy balance, as it receives and integrates signals from acylated ghrelin.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.63-1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Cell Lysates, Plasma, Serum, Tissue Homogenates
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias A-GHRLA-GHRL;
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 100 μL, Plasma 100 μL, Serum 100 μL, Tissue Homogenate 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Acylated Ghrelin
Test Time 1 hr 20 min
Total Assay Time 3 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.