Learn More
BIOHIPPO Human A-GHRL Microsample
SDP
Catalog No. 5000315807
Encompass
Human A-GHRL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of A-GHRL in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 9 24 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human A-GHRL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human A-GHRL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human A-GHRL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15807 Supplier BIOHIPPO Supplier No. BHE15210510
Human A-GHRL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of A-GHRL in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 9 24 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human A-GHRL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human A-GHRL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human A-GHRL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color