Learn More
American Research Products Inc Human A kinase anchor protein
Catalog No. 5000328538
Encompass
Human A-kinase anchor protein 4 (AKAP4) ELISA kit from CUSABIO Detection Range 25 pg/mL-1600 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28538 Supplier American Research Products Inc Supplier No. CSBEL001526HU
Human A-kinase anchor protein 4 (AKAP4) ELISA kit from CUSABIO Detection Range 25 pg/mL-1600 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich