Learn More
Sino Biological Anti-A1BG Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 207096T32100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human A1BG fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3868