Learn More
Sino Biological Human A2M PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10952T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human alpha-2-macroglobulin / A2M (rh alpha-2-macroglobulin / A2M; Catalog#10952-H08B; NP_000005.2; Met1-Ala1474). alpha-2-macroglobulin / A2M specific IgG was purified by Human alpha-2-macroglobulin / A2M affinity chromatography.
Catalog No. 50-237-2638