Learn More
Sino Biological Recombinant Human A2AR Protein (VLP, Full-length, Flag Tag)
Catalog No. 502993618
Encompass_Preferred
A DNA sequence encoding the Human AA2AR (P29274) (Met1-Ser412) was expressed with a flag tag at the N-terminus.
Catalog No. 50-299-3618 Supplier Sino Biological Supplier No. 12307H22H20
A DNA sequence encoding the Human AA2AR (P29274) (Met1-Ser412) was expressed with a flag tag at the N-terminus.