Learn More
Akron Biotechnology LLC Human AB Serum Closed System Solutions (CSS)TM Converted from Octaplas , Pooled Plasma (Human), Xeno-Free, Virus Inactivated, GMP Grade, 100 mL Bag, AR1048-0100
Supplier:  Akron Biotechnology LLC AR10480100
Encompass
Akron’s Human AB Serum, Converted from Octaplas, Pooled Plasma (Human), Xeno-Free, Virus Inactivated is manufactured, tested, and released following relevant cGMP guidelines for ancillary materials.Product Features: Plasma Source• Octaplas is an FDA-approved, sterile, pyrogen-free, frozen, S/D treated, prion-ligand treated, pooled human plasma• Solvent Detergent (S/D) treatment for inactivation of enveloped viruses• Immune Neutralization for inactivation of certain non-enveloped viruses• Affinity Chromatography intended to reduce prion proteinsSterile microfiltration minimizes the presence of bacteria and parasites.Stability• 24-month shelf life• Store at -20 °C• Transport on dry ice• Avoid repeated freeze-thaw cyclesFor Use StatementFor research use or further manufacturing use in ex vivo cell therapy applications. This product is not intended for direct in vivo use or for direct clinical use as a drug, therapeutic, biologic, or medical device.
Catalog No. NC2255755