Learn More
Bio IVT Human AB Serum, Plasma-Derived, RUO, Clear, straw yellow, Volume Options: 20 mL, 50 mL, 100 mL
Catalog No. NC3366058
Encompass
The product is human AB serum, a plasma-derived supplement that is essential for diverse cell culture applications. This serum is sourced from healthy male donors of the AB serotype to minimize immunoreactivity. It undergoes rigorous quality control testing to ensure its reliability in research applications, including cell expansion and gene therapy. The serum is defibrinated with bovine thrombin, providing optimal growth factors and nutrients for robust cell proliferation.
  • Reduced immunoreactivity from AB serotype donors
  • Optimized for cell culture applications
  • Consistent quality through stringent quality control
  • Thorough safety testing for sterility and viral markers
  • Flexible processing options available
Catalog No. NC3366058 Supplier Bio IVT Supplier No. HUMANABSRMP1
The product is human AB serum, a plasma-derived supplement that is essential for diverse cell culture applications. This serum is sourced from healthy male donors of the AB serotype to minimize immunoreactivity. It undergoes rigorous quality control testing to ensure its reliability in research applications, including cell expansion and gene therapy. The serum is defibrinated with bovine thrombin, providing optimal growth factors and nutrients for robust cell proliferation.
  • Reduced immunoreactivity from AB serotype donors
  • Optimized for cell culture applications
  • Consistent quality through stringent quality control
  • Thorough safety testing for sterility and viral markers
  • Flexible processing options available