Learn More
Abcam Human ABCA1 ELISA Kit, 1 x 96 Tests
Catalog No. 502655070
Encompass_Preferred
Human ABCA1 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human ABCA1 with a sensitivity of 0.06 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-5070 Supplier Abcam Supplier No. AB2846141X96TESTS
Human ABCA1 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human ABCA1 with a sensitivity of 0.06 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement