Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ABCA4 (aa 1166-1253) Control Fragment Recombinant Protein

Catalog No. RP109686
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP109686 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP109686 Supplier Invitrogen™ Supplier No. RP109686
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ABCA4 (ATP-Binding Cassette, Subfamily A, Member 4), also known as ABCR, is a protein which in humans is encoded by the ABCA4 gene. ABCA4 is a member of the ATP-binding cassette transporter gene sub-family A (ABC1) found exclusively in multicellular eukaryotes. Using a whole genome radiation hybrid panel, this gene is mapped to 1p21-p13. And this gene is expressed exclusively in retina photoreceptor cells, indicating the gene product mediates transport of an essental molecule across the photoreceptor cell membrane. Additionally, it is showed by immunofluorescence microscopy and Western blot analysis that ABCR is present in foveal and peripheral cone, as well as rod, photoreceptors. The results suggested that the loss in central vision experienced by patients with Stargardt macular dystrophy arises directly from ABCR-mediated foveal cone degeneration.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78363
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 24
Name Human ABCA4 (aa 1166-1253) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias ABC transporter; ABC10; abca4; abca4.L; Abcr; ARMD2; ATP binding cassette subfamily A member 4; ATP binding cassette subfamily A member 4 L homeolog; ATP binding cassette transporter; ATP-binding cassette 10; ATP-binding cassette sub-family A member 4; ATP-binding cassette transporter, retinal-specific; ATP-binding cassette, subfamily A (ABC1), member 4; ATP-binding cassette, sub-family A (ABC1), member 4; ATP-binding cassette, sub-family A, member 4; ATP-binding transporter, retina-specific; AW050280; CORD3; D430003I15Rik; FFM; photoreceptor rim protein; retinal ABCA4 transporter; retinal-specific ATP-binding cassette transporter; Retinal-specific phospholipid-transporting ATPase ABCA4; retina-specific ABC transporter; RIM ABC transporter; RIM protein; RMP; RP19; Stargardt disease protein; STGD; STGD1; XELAEV_180233141mg
Common Name ABCA4
Gene Symbol ABCA4
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IQSQRKGSEGTCSCSSKGFSTTCPAHVDDLTPEQVLDGDVNELMDVVLHHVPEAKLVECIGQELIFLLPNKNFKHRAYASLFRELEET
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.