Learn More
Sino Biological Anti-ABCD1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104898T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ABCD1, and purified by antigen affinity chromatography.
Catalog No. 50-240-8812