Learn More
Sino Biological Recombinant Human c-Abl/ABL1 Protein (GST Tag), HPLC-verified 500
Supplier:  Sino Biological 11199H09B500
Encompass_Preferred
A DNA sequence encoding the amino acid sequence (Pro 137-Ser 554) of human ABL1 isoform B (NP_009297.2) was fused with the GST tag at the N-terminus.
Catalog No. 50-224-6764