Learn More
Sino Biological Human ACBD6 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785766
Encompass_Preferred
The Human ACBD6 ELISA Pair Set is for the quantitative determination of Human ACBD6.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for ACBD6 coated on a 96-well plate. Standards and samples are added to the wells, and any ACBD6 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-ACBD6 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of ACBD6 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85766 Supplier Sino Biological Supplier No. SEK111125
The Human ACBD6 ELISA Pair Set is for the quantitative determination of Human ACBD6.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for ACBD6 coated on a 96-well plate. Standards and samples are added to the wells, and any ACBD6 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-ACBD6 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of ACBD6 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.