Learn More
Bon Opus Biosciences Recombinant Human Angiotensin-Converting Enzyme 2/ACE-2 (C-6His) 10ug
SDP
Supplier:  Bon Opus Biosciences C41910UG
Encompass_Preferred
Recombinant Human Angiotensin-Converting Enzyme 2 is produced by our Mammalian expression system and the target gene encoding Gln18-Ser740 is expressed with a 6His tag at the C-terminus. Accession# Q9BYF1. Formulation: Supplied as a 0.2 μm filtered solution of 20mM TrisHCl, 150mM NaCl, 0.1mM ZnCl2, 10% Glycerol, pH 7.5. Bon Opus Cat# C419
Catalog No. 50-217-2740