Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human ACE Quantikine ELISA Kit
SDP

Catalog No. DACE00
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DACE00 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DACE00 Supplier R&D Systems™ Supplier No. DACE00

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human ACE Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human ACE Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human ACE/CD143 in cell culture supernates (50 ul), serum (10 ul), heparin plasma (10 ul), saliva (25 ul).

Specifications

Assay Range 0.78 to 50 ng/mL
Assay Sensitivity 0.051 ng/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (Heparin), Saliva, Serum
Specificity Natural and Recombinant Human ACE. In addition to soluble ACE, this kit recognizes membrane-bound forms from lysates of cultured cells and supernates.
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 1636
Gene Symbol ACE
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),Heparin Plasma (10uL),Saliva (25uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA
Target ACE/CD143
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.