Learn More
ACROBiosystems ACROBiosystems Human ACE2 / ACEH Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems AC2H5255200UG
Encompass_Preferred
Recombinant Protein;200UG;Human ACE2, Fc Tag (AC2-H5255) is expressed from human 293 cells (HEK293).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 110.0 kDa. As a result of glycosylation, the protein migrates as 125-150 kDa under reducing (R) condition, and 250-300 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human ACE2, Fc Tag (Cat. No. AC2-H5255) at 2 μg/mL (100 μL/well) can bind Biotinylated SARS-CoV-2 S1 protein, His,Avitag (Cat. No. S1N-C82E8) with a linear range of 40-625 ng/mL (QC tested).
Catalog No. 50-210-7415